ENA429359113 Enamine

4-methylphthalazin-1-amine

  • MDL number MFCD00458086
  • Cas Number : 4776-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187121
Availability details for
Quantity
  • Availability
  • Add to Cart