ENA429359114 Enamine

(2S)-2-amino-5-ethoxy-5-oxopentanoic acid

  • MDL number MFCD00037217
  • Cas Number : 1119-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187122

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187122
Availability details for
Quantity
  • Availability
  • Add to Cart