ENA429359115 Enamine

propane-1-sulfinamide

  • MDL number MFCD28118599
  • Cas Number : 871939-84-3
  • Molecular Weight : 107.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187126
Availability details for
Quantity
  • Availability
  • Add to Cart