ENA429359117 Enamine

3-{[(tert-butoxy)carbonyl]amino}pyrrolidine-3-carboxylic acid

  • MDL number MFCD04115498
  • Cas Number : 869786-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187135
Availability details for
Quantity
  • Availability
  • Add to Cart