ENA429359120 Enamine

2,2-dimethylpropanethioic S-acid

  • MDL number MFCD00009729
  • Cas Number : 55561-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187146
Availability details for
Quantity
  • Availability
  • Add to Cart