ENA429359121 Enamine

[5-fluoro-2-(1H-1,2,4-triazol-1-yl)phenyl]methanol

  • MDL number MFCD11639940
  • Cas Number : 1096329-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187148
Availability details for
Quantity
  • Availability
  • Add to Cart