ENA429359122 Enamine

2-(1-cyanocyclobutyl)acetic acid

  • MDL number MFCD19231921
  • Cas Number : 1246210-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187151
Availability details for
Quantity
  • Availability
  • Add to Cart