ENA429359149 Enamine

6-ethylpyrimidine-2,4-dithiol

  • MDL number MFCD28125024
  • Cas Number : 52590-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187546
Availability details for
Quantity
  • Availability
  • Add to Cart