ENA429359152 Enamine

5-(chloromethyl)-2-(2,2,2-trifluoroethyl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD28125027
  • Cas Number : 1803607-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187935
Availability details for
Quantity
  • Availability
  • Add to Cart