ENA429359153 Enamine

tert-butyl 4-[4-(cyanosulfanyl)phenyl]piperazine-1-carboxylate

  • MDL number MFCD28125028
  • Cas Number : 1803607-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188283
Availability details for
Quantity
  • Availability
  • Add to Cart