ENA429359154 Enamine

1-(piperidin-4-yl)-1H-1,2,4-triazole-3-carbonitrile hydrochloride

  • MDL number MFCD28125029
  • Cas Number : 1803590-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188301
Availability details for
Quantity
  • Availability
  • Add to Cart