ENA429359157 Enamine

1,1-dioxo-1lambda6-thietane-3-sulfonyl chloride

  • MDL number MFCD28125032
  • Cas Number : 1803588-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188697
Availability details for
Quantity
  • Availability
  • Add to Cart