ENA429359158 Enamine

2-fluoro-1-(methoxymethoxy)-3-methylbenzene

  • MDL number MFCD28125033
  • Cas Number : 1698576-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188906
Availability details for
Quantity
  • Availability
  • Add to Cart