ENA429359167 Enamine

3-bromo-5-chloro-2-fluorobenzonitrile

  • MDL number MFCD11846061
  • Cas Number : 1160574-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187215
Availability details for
Quantity
  • Availability
  • Add to Cart