ENA429359168 Enamine

1-(5-methylpyridin-2-yl)piperidine-3-carboxylic acid

  • MDL number MFCD09741354
  • Cas Number : 954572-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187216
Availability details for
Quantity
  • Availability
  • Add to Cart