ENA429359169 Enamine

1-(2,2,2-trifluoroethyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD19550898
  • Cas Number : 1408390-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187217

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187217
Availability details for
Quantity
  • Availability
  • Add to Cart