ENA429359170 Enamine

4-(2-hydroxyethyl)-2,6-dimethoxyphenol

  • MDL number MFCD24714264
  • Cas Number : 20824-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187218

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187218
Availability details for
Quantity
  • Availability
  • Add to Cart