ENA429359176 Enamine

4-(cyclopropylmethoxy)benzene-1-carbothioamide

  • MDL number MFCD11136718
  • Cas Number : 1019568-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187227

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187227
Availability details for
Quantity
  • Availability
  • Add to Cart