ENA429359180 Enamine

N-(2,2-difluoroethyl)pyridin-4-amine hydrochloride

  • MDL number MFCD28118611
  • Cas Number : 1803588-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187231
Availability details for
Quantity
  • Availability
  • Add to Cart