ENA429359182 Enamine

(1r,3r)-1-bromo-3-ethoxycyclobutane

  • MDL number MFCD28118612
  • Cas Number : 59138-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187233
Availability details for
Quantity
  • Availability
  • Add to Cart