ENA429359183 Enamine

4-methyl-2-{[3-(trifluoromethyl)phenyl]methyl}-1,3-thiazole

  • MDL number MFCD26303545
  • Cas Number : 1350932-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187234
Availability details for
Quantity
  • Availability
  • Add to Cart