ENA429359192 Enamine

N-(5-fluoropyridin-2-yl)-1,3-thiazole-4-carboxamide

  • MDL number MFCD21375413
  • Cas Number : 1498694-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187246
Availability details for
Quantity
  • Availability
  • Add to Cart