ENA429359193 Enamine

N'-phenylpyridine-4-carbohydrazide

  • MDL number MFCD02344904
  • Cas Number : 91396-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187247

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-187247
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart