ENA429359194 Enamine

2,2-difluoro-2-(6-methylpyridin-3-yl)acetic acid hydrochloride

  • MDL number MFCD28118616
  • Cas Number : 1790141-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187248
Availability details for
Quantity
  • Availability
  • Add to Cart