ENA429359195 Enamine

2-cyclohexyl-4,5,6,7-tetrahydro-1H-1,3-benzodiazol-5-amine dihydrochloride

  • MDL number MFCD28118617
  • Cas Number : 1803584-04-4
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187251
Availability details for
Quantity
  • Availability
  • Add to Cart