ENA429359198 Enamine

potassium (azetidin-1-ylmethyl)trifluoroboranuide

  • MDL number MFCD18714969

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187255

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187255
Availability details for
Quantity
  • Availability
  • Add to Cart