ENA429359199 Enamine

methyl 2-[(ethylsulfanyl)methyl]benzoate

  • MDL number MFCD24341600
  • Cas Number : 157651-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187263
Availability details for
Quantity
  • Availability
  • Add to Cart