ENA429359201 Enamine

[(2,3-dimethylcyclohexyl)carbamoyl]methyl 2-hydroxy-3-methoxybenzoate

  • MDL number MFCD04751288
  • Cas Number : 1052594-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187266
Availability details for
Quantity
  • Availability
  • Add to Cart