ENA429359204 Enamine

2-bromopropanedinitrile

  • MDL number MFCD01570746
  • Cas Number : 1885-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187277

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187277
Availability details for
Quantity
  • Availability
  • Add to Cart