ENA429359208 Enamine

3-methyl-5-oxocyclohexane-1,2-dicarboxylic acid

  • MDL number MFCD24487753
  • Cas Number : 15941-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187287
Availability details for
Quantity
  • Availability
  • Add to Cart