ENA429359212 Enamine

4-(4-chloro-1H-pyrazol-1-yl)-3-fluorobenzonitrile

  • MDL number MFCD12779064
  • Cas Number : 1178197-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187301

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187301
Availability details for
Quantity
  • Availability
  • Add to Cart