ENA429359219 Enamine

6-(2-fluorophenyl)pyridazin-3-amine

  • MDL number MFCD12032762
  • Cas Number : 1159818-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187308
Availability details for
Quantity
  • Availability
  • Add to Cart