ENA429359220 Enamine

1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-4-ol

  • MDL number MFCD00109112
  • Cas Number : 1057670-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187309

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187309
Availability details for
Quantity
  • Availability
  • Add to Cart