ENA429359222 Enamine

1-(2-aminopyrimidin-4-yl)piperidin-3-ol

  • MDL number MFCD19543854
  • Cas Number : 1314355-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187311
Availability details for
Quantity
  • Availability
  • Add to Cart