ENA429359225 Enamine

1-methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD11209725
  • Cas Number : 92406-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187343

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187343
Availability details for
Quantity
  • Availability
  • Add to Cart