ENA429359227 Enamine

3-[(4-bromo-1H-pyrazol-1-yl)methyl]-5-propyl-1,2,4-oxadiazole

  • MDL number MFCD14679575
  • Cas Number : 1251048-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187346
Availability details for
Quantity
  • Availability
  • Add to Cart