ENA429359228 Enamine

{1-[(pyrimidin-2-yl)methyl]piperidin-4-yl}methanol

  • MDL number MFCD16848570
  • Cas Number : 138030-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187347
Availability details for
Quantity
  • Availability
  • Add to Cart