ENA429359229 Enamine

3-cyclopropylimidazolidin-4-one

  • MDL number MFCD26027280
  • Cas Number : 1535474-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187348
Availability details for
Quantity
  • Availability
  • Add to Cart