ENA429359231 Enamine

4-amino-N-(4,5-dihydro-1,3-thiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD01048678
  • Cas Number : 32365-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187350
Availability details for
Quantity
  • Availability
  • Add to Cart