ENA429359233 Enamine

2-(2-oxo-1,3-oxazinan-3-yl)benzonitrile

  • MDL number MFCD22720538
  • Cas Number : 1507014-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187353
Availability details for
Quantity
  • Availability
  • Add to Cart