ENA429359234 Enamine

[1-(4-fluoro-3-methylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD17227347
  • Cas Number : 1338949-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187354
Availability details for
Quantity
  • Availability
  • Add to Cart