ENA429359236 Enamine

3-cyclopentyl-1-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD26846195
  • Cas Number : 1565403-78-2
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187356
Availability details for
Quantity
  • Availability
  • Add to Cart