ENA429359243 Enamine

1-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]-1H-1,2,4-triazol-3-amine

  • MDL number MFCD12871960
  • Cas Number : 1178618-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187371
Availability details for
Quantity
  • Availability
  • Add to Cart