ENA429359246 Enamine

[1-(2,2-difluoroethyl)-1H-1,2,4-triazol-5-yl]methanol

  • MDL number MFCD21272699
  • Cas Number : 265643-98-9
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(2,2-difluoroethyl)-1H-1,2,4-triazol-5-yl]methanol
Inchi Code 1S/C5H7F2N3O/c6-4(7)1-10-5(2-11)8-3-9-10/h3-4,11H,1-2H2
Inchi Key UUHFGAPRERVWQL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187377
Availability details for
Quantity
  • Availability
  • Add to Cart