ENA429359247 Enamine

methyl 2-bromo-5-chloro-4-fluorobenzoate

  • MDL number MFCD24108550
  • Cas Number : 1502835-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187378

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-187378
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart