ENA429359248 Enamine

methyl 2,5-dimethyl-4-sulfanylfuran-3-carboxylate

  • MDL number MFCD26841991
  • Cas Number : 1565345-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187379
Availability details for
Quantity
  • Availability
  • Add to Cart