ENA429359249 Enamine

N-(2,2-difluoroethyl)-4-fluoroaniline

  • MDL number MFCD12807435
  • Cas Number : 1178500-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187381
Availability details for
Quantity
  • Availability
  • Add to Cart