ENA429359250 Enamine

2-benzylpropane-1,3-diol

  • MDL number MFCD00666386
  • Cas Number : 2612-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-70
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187382
Availability details for
Quantity
  • Availability
  • Add to Cart