ENA429359255 Enamine

5-(5-methylpyridin-3-yl)-1,2-oxazol-3-amine

  • MDL number MFCD21717330
  • Cas Number : 1494504-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187389
Availability details for
Quantity
  • Availability
  • Add to Cart