ENA429359257 Enamine

5-cyclopropyl-1,2-oxazol-3-amine

  • MDL number MFCD18379724
  • Cas Number : 21080-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187392

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-187392
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart